Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Myeloid differentiation primary response 88
- Antigen symbol:MYD88
- Clonality:Monoclonal
- Clone:2E9C2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:4615
- Antigen synonyms:MYD88D|myeloid differentiation primary response 88
- Amino acid number:176 to 280
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 176-280 of human MYD88.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant MYD88.
Recommended Dilutions: Western Blot: 2-4 mg/ml
Type: Primary
Antigen: MYD88
Clonality: Monoclonal
Clone: 2E9C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse