Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-C-peptide Mouse Monoclonal Antibody [clone: CII-55]
Anti-C-peptide Mouse Monoclonal Antibody [clone: CII-55]
Catalog # ABNOMAB2811
Supplier:  Abnova
Anti-C-peptide Mouse Monoclonal Antibody [clone: CII-55]
Catalog # ABNOMAB2811
Supplier:  Abnova

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C-peptide
  • Clonality:
    Monoclonal
  • Clone:
    CII-55
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Rat
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS, pH 7,4 (0,1% sodium azide)
  • Storage temperature:
    Store at 4 °C.
  • Immunogen:
    A synthetic peptide corresponding to rat C-peptide.
  • Size:
    200 μg
  • Pk:
    200 µg

Specifications

About this item

Mouse monoclonal antibody raised against synthetic peptide of C-peptide.

Type: Primary
Antigen: C-peptide
Clonality: Monoclonal
Clone: CII-55
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat