Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 2, Subfamily E, Polypeptide 1
- Antigen symbol:CYP2E1
- Clonality:Monoclonal
- Clone:1-98-1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:25086
- Antigen synonyms:CYPIIE1|Cytochrome P450 2E1|CYP2E|CYP2E1|4-nitrophenol 2-hydroxylase|Cytochrome P450-J
- Storage temperature:Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to rat Cyp2e1.
- Size:100 μl
- Pk:100 Tests
Mouse monoclonal antibody raised against recombinant Cyp2e1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: CYP2E1
Clonality: Monoclonal
Clone: 1-98-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Rat