Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:MEM-28
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:5788
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified PTPRC from human thymocytes and T lymphocytes.
- Size:100 Reactions
- Pk:100 Tests
Mouse monoclonal antibody raised against native PTPRC.
- Recommended Dilutions: Flow Cytometry: 10 μl in human blood cells 100 μl in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: MEM-28
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human