Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:CD8 antigen, alpha chain
- Antigen symbol:CD8A
- Clonality:Monoclonal
- Clone:MEM-31
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:925|926
- Antigen synonyms:p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CD8 from Crude thymus membrane fraction.
- Size:100 Reactions
- Pk:100 µl
Mouse monoclonal antibody raised against native CD8.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: MEM-31
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host:
Isotype: IgG2a
Reactivity: Human