Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:CD3 Molecule Delta
- Antigen symbol:CD3D
- Clonality:Monoclonal
- Clone:MEM-57
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:915|916|917
- Antigen synonyms:CD3-DELTA|IMD19|T3D
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CD3 from human thymocytes and T lymphocytes.
- Size:100 Reactions
- Pk:100 Tests
Specifications
About this item
Mouse monoclonal antibody raised against native CD3.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: CD3D
Clonality: Monoclonal
Clone: MEM-57
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human