Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Calcium Channel, Voltage-dependent, T type, alpha 1H subunit
- Antigen symbol:CACNA1H
- Clonality:Monoclonal
- Clone:S55-10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:8912
- Antigen synonyms:ECA6|EIG6|Calcium channel voltage-dependent T type alpha 1H subunit|Cav3.2|CACNA1HB
- Amino acid number:1019 to 1293
- Storage buffer:PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant fusion protein corresponding to amino acids 1019-1293 (II-III loop) of human CACNA1H.
- Size:100 μg
- Pk:100 µl
Mouse monoclonal antibody raised against partial recombinant CACNA1H.
Recommended Dilutions: Western Blot: 1-10 ?g/ml; Immunohistochemistry: 0.1-1.0 ?g/ml; Immunocytochemistry: 0.1-1.0 ?g/ml; Immunofluorescence: 1.0-10 ?g/ml
Type: Primary
Antigen: CACNA1H
Clonality: Monoclonal
Clone: S55-10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat