Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antigen name:Sterol regulatory element binding transcription factor 1
- Antigen symbol:SREBF1
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- Host:Rabbit
- Reactivity:Pig,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:6720
- Antigen synonyms:sterol regulatory element-binding protein 1,|SREBP1|SRBP1|SREBF1
- Amino acid number:300 to 400
- Storage buffer:50 mM sodium borate
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to amino acids 300-400 of human SREBF1.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of SREBF1.
Recommended Dilutions: Western Blot: 2 ?g/ml
Type:
Antigen: SREBF1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Pig, Rat