Anti-SIV type 1 Nef Mouse Monoclonal Antibody [clone: 17 (04-001)]
ABNOMAB7856
: Abnova
- Antibody type:Primary
- Antigen name:Simian immunodeficiency virus type 1 Nef
- Antigen symbol:SIV type 1 Nef
- Clonality:Monoclonal
- Clone:17 (04-001)
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Virus
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to Simian immunodeficiency virus type 1 (mac251 strain) Nef.
- Size:100 μl
- Pk:100 µg
Mouse monoclonal antibody raised against recombinant Simian immunodeficiency virus type 1 Nef.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000
Type: Primary
Antigen: SIV type 1 Nef
Clonality: Monoclonal
Clone: 17 (04-001)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus