Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Calcium Channel, Voltage-dependent, beta 2 subunit
- Antigen symbol:CACNB2
- Clonality:Monoclonal
- Clone:S8b-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:116600
- Antigen synonyms:CACNLB2|MYSB|CAVB2
- Amino acid number:189 to 205
- Storage buffer:PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 189-205 of rat Kcnt1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of Kcnt1.
Recommended Dilutions: Western Blot: 1-10 ?g/ml; Immunohistochemistry: 0.1-1.0 ?g/ml; Immunocytochemistry: 0.1-1.0 ?g/ml; Immunofluorescence: 1-10 ?g/ml
Type: Primary
Antigen: CACNB2
Clonality: Monoclonal
Clone: S8b-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat