Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-H2-D2 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 27-11-13S]
Anti-H2-D2 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 27-11-13S]
Catalog # ABNOMAB5823
Supplier:  Abnova
Anti-H2-D2 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 27-11-13S]
Catalog # ABNOMAB5823
Supplier:  Abnova

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    H2-D2
  • Clonality:
    Monoclonal
  • Clone:
    27-11-13S
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS (1 mg/mL sodium azide)
  • Storage temperature:
    Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    C3H alphaBDF1 splenocytes.
  • Size:
    500 μg
  • Pk:
    500 µg

Specifications

About this item

Mouse monoclonal antibody raised against H2-D2.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: H2-D2
Clonality: Monoclonal
Clone: 27-11-13S
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse