Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen symbol:Bu-1a
- Clonality:Monoclonal
- Clone:21-1A4
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Chicken
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (1 mg/mL sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from Bursal cells of one-day-old H.B15 (Bu-1 a/b) strain chickens.
- Size:500 μg
- Pk:500 µg
Specifications
About this item
Mouse monoclonal antibody raised against native Bu-1a.
Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells
Type: Primary
Antigen: Bu-1a
Clonality: Monoclonal
Clone: 21-1A4
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken