Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Farnesyl diphosphate synthase
- Antigen symbol:FDPS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:110196
- Antigen synonyms:FPPS|FPS
- Storage buffer:PBS (50% glycerol, 0,02% sodium azide)
- Sequence:FFQVQDDYLDLFGDPSVTGKVGTDIQDNKCSWLVVQCLLRASPQQRQILEENYGQKDPEKVARVKALYEALDLQSAFFKYEEDSYNRLKSLIEQCSAPLPPSIFMELANKIYKRRK
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant GST fusion protein corresponding to 116 amino acids of mouse Fdps.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Rabbit polyclonal antibody raised against partial recombinant Fdps.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: FDPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human