Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:X-ray repair complementing defective repair in Chinese hamster cells 5
- Antigen symbol:XRCC5
- Clonality:Monoclonal
- Clone:MEM-54
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:7520
- Antigen synonyms:NFIV|KUB2|Ku86|KARP1|KARP-1|thyroid-lupus autoantigen|Ku autoantigen|CTC85|Ku86 autoantigen related protein 1|nuclear factor IV|TLAA|DNA repair protein XRCC5|lupus Ku autoantigen protein p86|ATP-dependent DNA helicase 2 subunit 2|86 kDa subunit of Ku antigen|X-ray repair cross-complementing protein 5|CTCBF|ATP-dependent DNA helicase II 80 kDa subunit|KU80|CTC box-binding factor 85 kDa subunit|80kDa
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified XRCC5 from human whole T-lymphocytes.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against native XRCC5.
Recommended Dilutions: Immunocytochemistry (2 ?g/ml; Immunofluorescence Immunoprecipitation
Type: Primary
Antigen: XRCC5
Clonality: Monoclonal
Clone: MEM-54
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human