Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antigen name:Kinase Insert Domain Receptor
- Antigen symbol:KDR
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:16542
- Antigen synonyms:KDR|Kinase insert domain receptor|VEGFR-2|CD309|VEGFR2|FLK-1|Vascular endothelial growth factor receptor 2|Fetal liver kinase 1|FLK1|Protein-tyrosine kinase receptor flk-1
- Amino acid number:1200 to 1300
- Storage buffer:50 mM sodium borate
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to amino acids 1200-1300 of mouse Kdr.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Kdr.
Recommended Dilutions: Western Blot: 1:250-1:1000
Type:
Antigen: KDR
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human