Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-SYCP1 Rabbit Polyclonal Antibody (DyLight® 549)
Anti-SYCP1 Rabbit Polyclonal Antibody (DyLight® 549)
Catalog # ABNOPAB14959
Supplier:  Abnova
Anti-SYCP1 Rabbit Polyclonal Antibody (DyLight® 549)
Catalog # ABNOPAB14959
Supplier:  Abnova

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antigen name:
    Synaptonemal Complex Protein 1
  • Antigen symbol:
    SYCP1
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 549
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    6847
  • Antigen synonyms:
    SCP1|SCP-1|CT8|HOM-TES-14
  • Storage buffer:
    50 mM sodium borate
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Immunogen:
    A synthetic peptide corresponding to C-terminus of human SYCP1.
  • Size:
    100 μl
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of SYCP1.

Recommended Dilutions: Immunofluorescence: 1:300-1:750

Type:
Antigen: SYCP1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat