Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antigen name:Synaptonemal Complex Protein 1
- Antigen symbol:SYCP1
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:20957
- Antigen synonyms:SCP1|SCP-1|CT8|HOM-TES-14
- Storage buffer:50 mM sodium borate
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to C-terminus of mouse Sycp1.
- Size:100 μl
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of Sycp1.
Recommended Dilutions: Immunofluorescence: 1:100-1:750; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:750; Immunohistochemistry (Frozen sections): 1:750
Type:
Antigen: SYCP1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse