Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antigen name:Extracellular link Domain Containing 1
- Antigen symbol:LYVE1
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:114332
- Antigen synonyms:lymphatic vessel endothelial hyaluronic acid receptor 1|CRSBP-1|hyaluronic acid receptor|UNQ230/PRO263|extracellular link domain-containing 1|extracellular link domain-containing protein 1|extracellular link domain containing 1|cell surface retention sequence-binding protein 1|LYVE-1|HAR|cell surface retention sequence binding protein-1|XLKD1
- Amino acid number:250 to 318
- Storage buffer:50 mM sodium borate
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to amino acids 250-318 of mouse Lyve1.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Lyve1.
Recommended Dilutions: Flow Cytometry: 1:25
Type:
Antigen: LYVE1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse