Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antigen name:Transmembrane emp24 domain-containing protein 10
- Antigen symbol:TMED10
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Primates,Rabbit,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:10972
- Antigen synonyms:TMED10|Tmp 21 I|21 kDa transmembrane trafficking protein|Transmembrane protein Tmp21|TMED 10|Transmembrane emp24 domain containing protein 10|S31III125|p23|Transmembrane trafficking protein 21kD|S31I125|1110014C03Rik|Transmembrane emp24-like trafficking protein 10 yeast|MGC102351|p24delta|p24delta1|TMEDA_HUMAN.|p24 family protein delta 1|Tmp 21
- Amino acid number:100 to 200
- Storage buffer:50 mM sodium borate
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to amino acids 100-200 of human TMED10.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of TMED10.
Recommended Dilutions: Western Blot: 2 ?g/ml
Type:
Antigen: TMED10
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Primates, Rabbit, Rat