Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-PLIN3 Rabbit Polyclonal Antibody (DyLight® 549)
Anti-PLIN3 Rabbit Polyclonal Antibody (DyLight® 549)
Catalog # ABNOPAB15147
Supplier:  Abnova
Anti-PLIN3 Rabbit Polyclonal Antibody (DyLight® 549)
Catalog # ABNOPAB15147
Supplier:  Abnova

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antigen name:
    Perilipin 3
  • Antigen symbol:
    PLIN3
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 549
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    66905
  • Antigen synonyms:
    1300012C15Rik|M6prbp1|Tip47
  • Amino acid number:
    1 to 100
  • Storage buffer:
    50 mM sodium borate
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Immunogen:
    A synthetic peptide corresponding to amino acids 1-100 of mouse Plin3.
  • Size:
    100 μl
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of Plin3.

Recommended Dilutions: Western Blot: 1:1000-1:5000

Type:
Antigen: PLIN3
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse