Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antigen name:Baculoviral IAP Repeat Containing 5
- Antigen symbol:BIRC5
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:332
- Antigen synonyms:IAP4|Survivin|API4|Apoptosis inhibitor 4|Baculoviral IAP repeat-containing protein 5|EPR-1
- Modification:phospho-Ser20
- Storage buffer:50 mM sodium borate
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S20 of human BIRC5.
- Size:100 μl
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic phosphopeptide of BIRC5.
Recommended Dilutions: Western Blot: 3 ?g/ml
Type:
Antigen: BIRC5
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human