Anti-17-beta-19-Nortestosterone Polyclonal Antibody
ABNOPAB8205
: Abnova
- Antibody type:Primary
- Antigen name:17-beta-19-Nortestosterone
- Antigen symbol:17-beta-19-Nortestosterone
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Cross adsorption:No
- Format:Liquid
- Storage buffer:20 mM phosphate buffer, 150 mM NaCl, pH 7,2 (0,09% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:17 beta-19-Nortestosterone conjugated with BTG.
- Pk:1 mg
Sheep polyclonal antibody raised against 17 beta-19-Nortestosterone.
Recommended Dilutions: Enzyme Immunoassay (10 ?g/ml
Type: Primary
Antigen: 17-beta-19-Nortestosterone
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: