Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-GRIN2C Rabbit Polyclonal Antibody
Anti-GRIN2C Rabbit Polyclonal Antibody
  ABGEAP19779C
 :  Abgent
Anti-GRIN2C Rabbit Polyclonal Antibody
  ABGEAP19779C
 :  Abgent

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate receptor, ionotropic, N-methyl D-aspartate 2C
  • Antigen symbol:
    GRIN2C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    2905
  • Antigen synonyms:
    NMDAR2C|GluN2C|NR2C
  • Amino acid number:
    567 to 595
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    134.209 kDa
  • Sequence:
    YNQNLTRGKKSGGPC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.32
  • Shipping temperature:
    4 °C
  • Immunogen:
    This GRIN2C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 567-595 amino acids from the Central region of human GRIN2C.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: GRIN2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Mouse