Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-IgG Chicken Polyclonal Antibody (CF™ 350)
Anti-IgG Chicken Polyclonal Antibody (CF™ 350)
Catalog # 20364-500UL
Supplier:  Biotium
Anti-IgG Chicken Polyclonal Antibody (CF™ 350)
Catalog # 20364-500UL
Supplier:  Biotium

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    CF® 350
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Chicken
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgY
  • Reactivity:
    Goat
  • Western blot:
    Yes
  • Size:
    500 μL
  • Pk:
    0,5 mL

Specifications

About this item

CF™350 is a blue fluorescent dye with excitation/emission maxima at 347/448nm. CF™350 is optimized for UV excitation, and is spectrally similar to AlexaFluor®350, AMCA, and DyLight®350. CF™350 is more water soluble, more photostable, and 50% brighter when conjugated to antibodies than the traditional blue dye AMCA. CF™350 antibody conjugates are comparable in brightness to AlexaFluor®350 dye conjugates.

CF™ dyes are covered by pending U.S. and international patents. CF™ is a trademark of Biotium, Inc. Alexa Fluor® is a registered trademark of Molecular Probes, Inc. DyLight® is a registered trademark of Pierce Biotechnology.

Ordering information: Antibodies are supplied at 2mg/mL in PBS pH 7.4 containing 50% glycerol, 2mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. Highly cross-adsorbed antibodies are adsorbed against whole serum protein and have minimal cross-reactivity for multiple labeling experiments as indicated. Bv: bovine; Ch: chicken; Gt: goat; GP: Guinea pig; Hs: horse; Hu: human; Ms: mouse; Rb: rabbit; Sh: sheep; SHm: Syrian hamster; Sw: swine; Rt: rat.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF350
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Goat