Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:CF® 568
- ELISA:Yes
- Flow cytometry:Yes
- Host:Chicken
- ImmunoFluorescence:Yes
- Isotype:IgY
- Reactivity:Mouse
- Western blot:Yes
- Size:50 μl
- Pk:50 µl
CF™568 antibodies are affinity-purified antibodies labeled with red fluorescent CF™568, a member of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™568 is a red fluorescent dye optimized for the 568nm laser line. CF™568 is spectrally similar to AlexaFluor®568, ATTO 565, and Rhodamine Red. Antibody conjugates of CF™568 are significantly brighter and more photostable than those of AlexaFluor®568, making CF™568 a better choice for demanding applications such as confocal microscopy.
CF™ dyes are covered by pending U.S. and international patents. CF™ is a trademark of Biotium, Inc. Alexa Fluor® is a registered trademark of Molecular Probes, Inc.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF568
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Mouse