Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:CF® 543
- ELISA:Yes
- Flow cytometry:Yes
- Host:Chicken
- ImmunoFluorescence:Yes
- Isotype:IgY
- Reactivity:Rabbit
- Western blot:Yes
- Size:50 μl
- Pk:50 µl
Specifications
About this item
CF™543 antibodies are affinity-purified antibodies labeled with orange fluorescent CF™543 (Ex/Em 541/560nm), a member of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™543 is an orange fluorescent dye with an excitation spectrum optimally matching the 543 laser line. Antibody conjugates of the dye are much brighter than those of Alexa Fluor® 546.
CF™ dyes are covered by pending U.S. and international patents.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF543
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Rabbit