Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 549)
ROCK609-142-002-05
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- Host:Goat
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Size:500 µg
- Pk:500 µG
Type: Secondary
Antigen: IgG(Human)-HL
Clonality: Polyclonal
Clone:
Conjugation: DyLight549
Epitope:
Host: Goat
Isotype:
Reactivity: Human