Anti-GRIN1 Mouse Monoclonal Antibody (Unconjugated) [clone: R1JHL]
ROCK900-301-C85
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, N-methyl D-aspartate 1
- Antigen symbol:GRIN1
- Clonality:Monoclonal
- Clone:R1JHL
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Cell Culture Supernatant
- Form:Lyophilized
- Gene ID:24408
- Antigen synonyms:NMDAR1|GluN1|NR1
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Anti-NMDA NR1 Subunit Antibody was produced by repeated immunizations with a fusion Proteins containing amino acids 1-564 of the rat NR1 subunit.
- Size:15 µg
- Pk:15 µG
Anti-NMDA NR1 Subunit (Mouse) antibody is suitable for use in Western Blotting and Immunoprecipitation. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 120 kDa in size corresponding to NR1 subunit.
Recommended Dilutions: Western Blot: 1:1000; Immunoprecipitation: 3µg per 200µg lysate;
Type: Primary
Antigen: GRIN1
Clonality: Monoclonal
Clone: R1JHL
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse, Rat