Anti-GRIN1 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-D87
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, N-methyl D-aspartate 1
- Antigen symbol:GRIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:24408
- Antigen synonyms:NMDAR1|GluN1|NR1
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Anti-NMDA R1, Splice Variant C2' Antibody was produced by repeated immunizations with synthetic peptide corresponding to amino acid residues specific to the NR1 subunit, C2' splice variant insert.
- Purification:affinity-purified
- Size:25 µg
- Pk:25 µG
Specifications
About this item
Anti-NMDA R1, Splice Variant C2' (Rabbit) Antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 120 kDa in size corresponding to the NMDA receptor.
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:1000-2000;
0.05 M Tris Chloride, 0.15 M Sodium Chloride, pH 7.4;
Type: Primary
Antigen: GRIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat