Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 549)
Catalog # ROCK603-142-002-05
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- Host:Goat
- ImmunoFluorescence:Yes
- Reactivity:Golden syrian hamster,Armenian hamster
- Western blot:Yes
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Secondary
Antigen: IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken,