Anti-VSV-G Rabbit Polyclonal Antibody (DyLight® 549)
Catalog # ROCK600-442-386
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Tag
- Antigen symbol:VSV-G
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Multiple
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Tag
Antigen: VSV-G
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan