Anti-HCN1 Mouse Monoclonal Antibody (Unconjugated) [clone: S70-28]
Catalog # ROCK200-301-F40
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:hyperpolarization-activated cyclic nucleotide-gated potassium channel 1
- Antigen symbol:HCN1
- Clonality:Monoclonal
- Clone:S70-28
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:84390
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
- Storage temperature:–20 °C
- Concentration:1 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:HCN1 Antibody was produced in mice by repeated immunizations with a fusion Protein corresponding to the C terminus region of rat HCN1.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-HCN1 Antibody is suitable for use in WB, IP and IHC. Expect a band approximately ~100kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: HCN1
Clonality: Monoclonal
Clone: S70-28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat