Anti-CACNA1H Mouse Monoclonal Antibody (Unconjugated) [clone: S55-10]
Catalog # ROCK200-301-F11
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:calcium channel, voltage-dependent, T type, alpha 1H subunit
- Antigen symbol:CACNA1H
- Clonality:Monoclonal
- Clone:S55-10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:Internal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:8912
- Antigen synonyms:ECA6|EIG6|Cav3.2|CACNA1HB
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:Cav3.2 Antibody was produced in mice by repeated immunizations with a fusion Protein corresponding to an internal region (II-III loop) of human Cav3.2.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-CAV3.2 Antibody is suitable for use in WB, IP and Immunofluorescence microscopy. Expect a band approximately ~260kDa on specific lysates. Specific conditions for reactivity should be optimised by the end user.
Type: Primary
Antigen: CACNA1H
Clonality: Monoclonal
Clone: S55-10
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat, Human