Anti-ACK Mouse Polyclonal Antibody (Unconjugated) [clone: 7F8]
Catalog # ROCK200-301-A97
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Acetyl-Lysine
- Antigen symbol:AcK
- Clonality:Polyclonal
- Clone:7F8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Birds,Mammals
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,09% (w/v) Sodium Azide, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This Protein G purified monoclonal antibody was prepared using conventional hybridoma technology after repeated immunizations with acetylated keyhole limpet hemocyanine (KLH).
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This Protein G purified antibody has been tested for use in ELISA, immunochemistry and western blotting. Specific conditions for reactivity should be optimized by the end user. Approximately 1 µg of antibody is sufficient to detect 20 µg.
Recommended Dilutions: Western Blot: 1:500 - 1:2000; Immunochemistry: 1:200 - 1:1000;
Type: Primary
Antigen: ACK
Clonality: Polyclonal
Clone: 7F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mammalian, Avian,