Anti-IgG F(c) Goat Polyclonal Antibody (DyLight® 549)
Catalog # ROCK611-142-003
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG F(c)
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- Host:Goat
- ImmunoFluorescence:Yes
- Reactivity:Rabbit
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Rabbit IgG F(c) fragment
- Tested applications:Fluoresence ELISA
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope: F(c)
Host: Goat
Isotype:
Reactivity: Rabbit