Anti-GRIN2A Rabbit Polyclonal Antibody
ROCK612-401-D89
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, N-methyl D-aspartate 2A
- Antigen symbol:GRIN2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:24409
- Antigen synonyms:NR2A|GluN2A|NMDAR2A
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-NMDA R2A Antibody was produced by repeated immunizations with a synthetic peptide corresponding to amino acid residues from the N-terminal region of the NR2A subunit.
- Purification:affinity-purified
- Size:100 µl
- Pk:100 µl
Anti-NMDA R2A Antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 180 kDa in size corresponding to the NR2A subuint of the NMDA receptor.
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:500;
Type: Primary
Antigen: GRIN2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Rat