Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog # ROCK612-401-C88
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, N-methyl D-aspartate 2B
- Antigen symbol:GRIN2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Rat
- Western blot:Yes
- ChIP:Yes
- Gene ID:24410
- Antigen synonyms:GluN2B
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Anti-NMDA NR2B Subunit pY1336 (Rabbit) antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 180 kDa in size corresponding to NMDAR NR2B subunit protein.
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:400;
Type: Primary
Antigen: GRIN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat