Anti-GST Goat Polyclonal Antibody (DyLight® 549)
Catalog # ROCK600-142-200
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Glutathione-S-Transferase
- Antigen symbol:GST
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Multiple
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Goat
Isotype:
Reactivity: pan