Anti-6xHis Rabbit Polyclonal Antibody (DyLight® 549)
ROCK600-442-382
- Antibody type:Tag
- Antigen name:His Tag
- Antigen symbol:6xHis
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Type: Tag
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan