Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Platelet/endothelial cell adhesion molecule 1
- Antigen symbol:PECAM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
- Storage temperature:–20 °C
- Concentration:200 ug/ml
- Immunogen:PECAM1 (Val421~Lys629); MGHHHHHHSGSEF- VIKGQTIEVR CESISGTLPI SYQLLKTSKV LENSTKNSND PAVFKDNPTE DVEYQCVADN CHSHAKMLSE VLRVKVIAPV DEVQISILSS KVVESGEDIV LQCAVNEGSG PITYKFYREK EGKPFYQMTS NATQAFWTKQ KASKEQEGEY YCTAFNRANH ASSVPRSKIL TVRVILAPWK KGLIAVVIIG VIIALLIIAA KCYFLRKAK
- Purification:purified by immunoaffinity chromatography
- Size:50 μg
- Pk:50 µG
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: Platelet/Endothelial Cell Adhesion Molecule
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,