Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Platelet Factor 4 Rabbit Polyclonal Antibody
Anti-Platelet Factor 4 Rabbit Polyclonal Antibody
Catalog # USBI141908
Supplier:  US Biological
Anti-Platelet Factor 4 Rabbit Polyclonal Antibody
Catalog # USBI141908
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Platelet Factor 4
  • Antigen symbol:
    PF4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Amino acid number:
    30 to 105
  • Storage buffer:
    PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
  • Sequence:
    MGSSHHHHHHSSGLVPRGSHMASMTGGQQMGRGSEFVTSAGPEESDGDLSCVCVKTISSGIHLKHITSLEVIKAGRHCAVPQLIATLKNGRKICLDRQAPLYKKVIKKIS
  • Storage temperature:
    –20 °C
  • Concentration:
    200 ug/ml
  • Immunogen:
    PF4 (Val30~Ser105); MGSSHHHHHHSSGLVPRGSHMASMTGGQQMGRGSEF-V TSAGPEESDG DLSCVCVKTI SSGIHLKHIT SLEVIKAGRH CAVPQLIATL KNGRKICLDR QAPLYKKVIK KILES
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: Platelet Factor 4
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse,