Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Amyloid Beta, 1-40 Rabbit Polyclonal Antibody
Anti-Amyloid Beta, 1-40 Rabbit Polyclonal Antibody
Catalog # USBI139198
Supplier:  US Biological
Anti-Amyloid Beta, 1-40 Rabbit Polyclonal Antibody
Catalog # USBI139198
Supplier:  US Biological

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Amyloid beta, 1-40
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
  • Storage temperature:
    –20 °C
  • Concentration:
    200 ug/ml
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: Amyloid Beta, 1-40
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat,