Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast Growth Factor, Basic
- Antigen symbol:FGF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Pig,Human,Bovine
- Western blot:Yes
- Cross adsorption:No
- Format:Serum
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2
- Storage temperature:–20 °C
- Immunogen:Recombinant bovine Fibroblast Growth Factor-2 [FGF-2] (basic FGF)
- Tested applications:Radio Immunoassay
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Immunochemistry: 1:1200-5000; Western Blot: 1:10000-1:30000
Type: Primary
Antigen: Fibroblast Growth Factor, Basic
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Pig,