Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Atrial Natriuretic Peptide, pro-
- Clonality:Monoclonal
- Clone:A137-M6-19-B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3 kappa
- Cross adsorption:No
- Form:lyophilized powder
- Amino acid number:40 to 55
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Synthetic peptide corresponding to aa40-55, (C)EPNEEAGAALSPLPEV of human pro-ANP.
- Purification:purified by Protein G chromatography
- Size:10 μg
- Pk:10 µG
Type: Primary
Antigen: Atrial Natriuretic Peptide, pro-
Clonality: Monoclonal
Clone: A137-M6-19-B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: