Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-VIM Rabbit Polyclonal Antibody
Anti-VIM Rabbit Polyclonal Antibody
Catalog # USBIV2122-01F
Supplier:  US Biological
Anti-VIM Rabbit Polyclonal Antibody
Catalog # USBIV2122-01F
Supplier:  US Biological

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Vimentin
  • Antigen symbol:
    VIM
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    lyophilized powder
  • Amino acid number:
    70 to 120
  • Storage buffer:
    PBS, pH 7,4, 0,02% sodium azide.
  • Storage temperature:
    –20 °C
  • Concentration:
    ~0,5 mg/ml
  • Immunogen:
    Synthetic peptide corresponding to a region within residues 70-120 of human vimentin, conjugated to KLH (Swiss Prot: P08670).
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    40 µg
  • Pk:
    40 µG

Specifications

About this item