Anti-Cre Recombinase Mouse Monoclonal Antibody [clone: 9E2]
Catalog # USBIC7920-01
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cre Recombinase
- Clonality:Monoclonal
- Clone:9E2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Western blot:Yes
- Cross adsorption:No
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,4, 1% BSA, 0,02% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,5 mg/ml (after reconstitution)
- Immunogen:Purified recombinant Cre recombinase protein
- Purification:purified by Protein A chromatography
- Size:40 µg
- Pk:40 µG