Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Splicing Factor 4
- Antigen symbol:SF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rooster,Human
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide, before the addition of glycerol to 40%.
- Sequence:TDAPTSAPSAPPS
- Storage temperature:–20 °C
- Concentration:~0,1 mg/ml
- Immunogen:Synthetic peptide, TDAPTSAPSAPPS, corresponding to human Splicing Factor 4 (KLH).
- Purification:purified by immunoaffinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:62500
Type: Primary
Antigen: Splicing Factor 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rooster,dent,