Inspection date labels, tamper-proof
Supplier: Brady
Certificates
About this item
These inspection date labels are made of polyolefin with printed German language.
- Hole pliers can be used for identification of the test date
- Tamper-proof: tears easily, can not be replaced as a whole
The labels can be used wherever regular checks on equipment and machinery are needed and also for fire-fighting equipment and more.