Anti-Dehydrin Rabbit Polyclonal Antibody
Catalog # AGRIAS07206
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:Dehydrin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana,N. tabacum
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated peptide sequence (K-segment) from dehydrin C terminal conserved in a wide range of plant species including Nicotiana tabacum BAD1349
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: Dehydrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, N. tabacum