Anti-Slc6a9 Rabbit Polyclonal Antibody
Catalog # ORIGTA328860
Supplier: OriGene
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen symbol:Solute carrier family 6, member 9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:202 to 214
- Storage buffer:Lyophilized. Concentration before lyophilization ~0,8mg/ml (lot dependent, please refer to CoA along with shipment for actual concentration). Buffer before lyophilization: Phosphate buffered saline (PBS), pH 7,4, 1% BSA, 0,05% NaN3.
- Sequence:C-RLYVLKLSDDIGD
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Peptide (C)RLYVLKLSDDIGD, corresponding to amino acid residues 202-214 of rat Glycine Transporter 1. 2nd extracellular loop.
- Purification:purified by antigen affinity chromatography
- Size:200 μl
- Pk:200 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:200-1:2000
Type: Primary
Antigen: Slc6a9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat